ENA413170987 Enamine

3-amino-2-[(3,4-difluorophenyl)methyl]propan-1-ol

  • MDL number MFCD11126839
  • Cas Number : 1019116-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154365
Availability details for
Quantity
  • Availability
  • Add to Cart