ENA413170988 Enamine

5-amino-2-phenoxybenzonitrile

  • MDL number MFCD14617347
  • Cas Number : 288252-04-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154368

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154368
Availability details for
Quantity
  • Availability
  • Add to Cart