ENA413170990 Enamine

1-(2-methoxyethyl)azetidin-3-ol

  • MDL number MFCD18308954
  • Cas Number : 1340368-69-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154372
Availability details for
Quantity
  • Availability
  • Add to Cart