ENA413170994 Enamine

1-(1-methoxypropan-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD19654226
  • Cas Number : 1341630-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154388
Availability details for
Quantity
  • Availability
  • Add to Cart