ENA413171002 Enamine

5-(methylsulfanyl)-1,3,4-thiadiazole-2-carbonitrile

  • MDL number MFCD18805205
  • Cas Number : 70391-06-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154465

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154465
Availability details for
Quantity
  • Availability
  • Add to Cart