ENA413171004 Enamine

3-(butan-2-yloxy)-N-methylaniline

  • MDL number MFCD10687536
  • Cas Number : 1040686-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154470

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154470
Availability details for
Quantity
  • Availability
  • Add to Cart