ENA413171005 Enamine

[1-(2,4-difluorophenyl)cyclopentyl]methanamine

  • MDL number MFCD20362010
  • Cas Number : 1341830-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154471

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154471
Availability details for
Quantity
  • Availability
  • Add to Cart