ENA413171006 Enamine

5-(2,2-dimethylmorpholine-4-carbonyl)-1H-pyrazol-3-amine

  • MDL number MFCD20316362
  • Cas Number : 1347108-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154481

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154481
Availability details for
Quantity
  • Availability
  • Add to Cart