ENA413171009 Enamine

2-[(methylsulfanyl)methyl]oxirane

  • MDL number MFCD08703703
  • Cas Number : 45378-62-9
  • Molecular Weight : 104.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-154484

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154484
Availability details for
Quantity
  • Availability
  • Add to Cart