ENA413171015 Enamine

4,4-diethylpiperidine hydrochloride

  • MDL number MFCD27500918
  • Cas Number : 3970-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154506

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154506
Availability details for
Quantity
  • Availability
  • Add to Cart