ENA413171026 Enamine

4-amino-N,3-dimethyl-1H-pyrazole-5-carboxamide

  • MDL number MFCD16669732
  • Cas Number : 68375-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154559

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154559
Availability details for
Quantity
  • Availability
  • Add to Cart