ENA413171028 Enamine

(1R)-1-(2,5-dichlorophenyl)ethan-1-amine hydrochloride

  • MDL number MFCD12757833
  • Cas Number : 1152088-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154565

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154565
Availability details for
Quantity
  • Availability
  • Add to Cart