ENA413171045 Enamine

6-(4-fluorophenyl)-6-oxohexanoic acid

  • MDL number MFCD01311599
  • Cas Number : 343319-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154611

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154611
Availability details for
Quantity
  • Availability
  • Add to Cart