ENA413171054 Enamine

3-(2-methoxyphenyl)-4,5-dihydro-1,2-oxazol-5-one

  • MDL number MFCD09751665
  • Cas Number : 24031-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-154633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154633
Availability details for
Quantity
  • Availability
  • Add to Cart