ENA413171060 Enamine

4,4-dimethyl-4H,5H,6H-cyclopenta[d][1,3]thiazol-2-amine

  • MDL number MFCD12923025
  • Cas Number : 1182284-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154685

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154685
Availability details for
Quantity
  • Availability
  • Add to Cart