ENA413171066 Enamine

4-amino-2,3,5-trimethylphenol

  • MDL number MFCD09833890
  • Cas Number : 10486-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154699
Availability details for
Quantity
  • Availability
  • Add to Cart