ENA413171089 Enamine

2-(ethylsulfamoyl)benzene-1-sulfonyl chloride

  • MDL number MFCD16041881
  • Cas Number : 1250663-44-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154745

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154745
Availability details for
Quantity
  • Availability
  • Add to Cart