ENA413171097 Enamine

(3S)-1-(4-nitrophenyl)pyrrolidin-3-ol

  • MDL number MFCD11037808
  • Cas Number : 791850-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154804

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154804
Availability details for
Quantity
  • Availability
  • Add to Cart