ENA413171101 Enamine

2-fluoro-2-phenylethan-1-amine

  • MDL number MFCD19204011
  • Cas Number : 55601-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152199
Availability details for
Quantity
  • Availability
  • Add to Cart