ENA413171102 Enamine

2-[4-(trifluoromethyl)phenyl]-1H-imidazole-5-carboxylic acid

  • MDL number MFCD07378851
  • Cas Number : 328569-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 229-230
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152207
Availability details for
Quantity
  • Availability
  • Add to Cart