ENA413171103 Enamine

5-bromo-3,7-dimethyl-1-benzofuran-2-carboxylic acid

  • MDL number MFCD18293973
  • Cas Number : 1343238-39-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152211

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152211
Availability details for
Quantity
  • Availability
  • Add to Cart