ENA413171105 Enamine

methyl 4-bromo-2-methoxy-5-nitrobenzoate

  • MDL number MFCD24189634
  • Cas Number : 98590-43-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154850

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154850
Availability details for
Quantity
  • Availability
  • Add to Cart