ENA413171110 Enamine

4-(aminomethyl)-1-(4-chlorophenyl)pyrrolidin-2-one

  • MDL number MFCD11579982
  • Cas Number : 1082524-78-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154856

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154856
Availability details for
Quantity
  • Availability
  • Add to Cart