ENA413171111 Enamine

1-(4-bromo-2-methoxyphenyl)methanamine

  • MDL number MFCD11045219
  • Cas Number : 1071037-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154857
Availability details for
Quantity
  • Availability
  • Add to Cart