ENA413171112 Enamine

(3-chloro-4-fluorophenyl)(1-methyl-1H-pyrazol-4-yl)methanamine

  • MDL number MFCD17291138
  • Cas Number : 1343711-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154858

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154858
Availability details for
Quantity
  • Availability
  • Add to Cart