ENA413171113 Enamine

2-phenylpent-4-enoic acid

  • MDL number MFCD20638191
  • Cas Number : 94086-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-152224

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-152224
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart