ENA413171116 Enamine

2-(4-methylbenzoyl)-1H-indol-3-amine

  • MDL number MFCD27500943
  • Cas Number : 1552304-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154869
Availability details for
Quantity
  • Availability
  • Add to Cart