ENA413171118 Enamine

3-(6-chloropyridazin-3-yl)benzonitrile

  • MDL number MFCD19560044
  • Cas Number : 99708-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154881

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154881
Availability details for
Quantity
  • Availability
  • Add to Cart