ENA413171120 Enamine

2-(1-cyano-1-methylethyl)-6-fluorobenzonitrile

  • MDL number MFCD19695188
  • Cas Number : 1314783-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154883

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-154883
Availability details for
Quantity
  • Availability
  • Add to Cart