ENA413171358 Enamine

methyl 5-({[(tert-butoxy)carbonyl]amino}methyl)-4,5-dihydro-1,2-oxazole-3-carboxylate

  • MDL number MFCD27920211
  • Cas Number : 1797234-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-154967

Safety Information

msds https://www.enaminestore.com/catalog/EN300-154967
Availability details for
Quantity
  • Availability
  • Add to Cart