ENA413171364 Enamine

1,2-dihydrospiro[indole-3,3'-pyrrolidine]-2-one hydrochloride

  • MDL number MFCD27500955
  • Cas Number : 1803592-37-1
  • Molecular Weight : 224.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name spiro[indoline-3,3'-pyrrolidin]-2-one hydrochloride
Inchi Code 1S/C11H12N2O.ClH/c14-10-11(5-6-12-7-11)8-3-1-2-4-9(8)13-10;/h1-4,12H,5-7H2,(H,13,14);1H
Inchi Key GEZIWWQRNMVBDS-UHFFFAOYSA-N

Description

Purity 95
Salt Data Cl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-155044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155044
Availability details for
Quantity
  • Availability
  • Add to Cart