ENA413171365 Enamine

4-methyl-4-(propan-2-yl)piperidine hydrochloride

  • MDL number MFCD27500956
  • Cas Number : 1123-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155087

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155087
Availability details for
Quantity
  • Availability
  • Add to Cart