ENA413171373 Enamine

7-ethoxy-2,3-dihydro-1,4-benzodioxin-6-amine hydrochloride

  • MDL number MFCD27500964
  • Cas Number : 1797553-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155126

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155126
Availability details for
Quantity
  • Availability
  • Add to Cart