ENA413171374 Enamine

2-(chloromethyl)-5,6-dimethoxy-1H-1,3-benzodiazole hydrochloride

  • MDL number MFCD27500965
  • Cas Number : 1803582-86-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155129

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155129
Availability details for
Quantity
  • Availability
  • Add to Cart