ENA413171375 Enamine

2-azaspiro[3.4]octane hydrochloride

  • MDL number MFCD27500966
  • Cas Number : 1414885-15-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155135
Availability details for
Quantity
  • Availability
  • Add to Cart