ENA413171376 Enamine

benzyl[3-(difluoromethoxy)propyl]amine

  • MDL number MFCD27920213
  • Cas Number : 1593441-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155139
Availability details for
Quantity
  • Availability
  • Add to Cart