ENA413171384 Enamine

5-fluoro-9-oxo-9,10-dihydroacridine-4-carboxylic acid

  • MDL number MFCD27920216
  • Cas Number : 98370-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155315

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155315
Availability details for
Quantity
  • Availability
  • Add to Cart