ENA413171385 Enamine

2,5-difluoro-9-oxo-9,10-dihydroacridine-4-carboxylic acid

  • MDL number MFCD27920217
  • Cas Number : 1797691-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155317
Availability details for
Quantity
  • Availability
  • Add to Cart