ENA413171386 Enamine

2,7-difluoro-9-oxo-9,10-dihydroacridine-4-carboxylic acid

  • MDL number MFCD27920218
  • Cas Number : 1797801-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155318

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155318
Availability details for
Quantity
  • Availability
  • Add to Cart