ENA413171387 Enamine

2-(4-methylbenzoyl)furan-3-amine

  • MDL number MFCD27920219
  • Cas Number : 1803592-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155322
Availability details for
Quantity
  • Availability
  • Add to Cart