ENA413171389 Enamine

5-methyl-N-(propan-2-yl)-4H-1,2,4-triazol-3-amine

  • MDL number MFCD27500975
  • Cas Number : 1803600-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-155376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155376
Availability details for
Quantity
  • Availability
  • Add to Cart