ENA413171402 Enamine

4-[(4-bromophenyl)methoxy]phenyl benzoate

  • MDL number MFCD27920222
  • Cas Number : 1803582-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155482

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155482
Availability details for
Quantity
  • Availability
  • Add to Cart