ENA413171403 Enamine

methyl 5-carbamoylfuran-3-carboxylate

  • MDL number MFCD27920223
  • Cas Number : 1803561-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155486

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155486
Availability details for
Quantity
  • Availability
  • Add to Cart