ENA413171405 Enamine

4-(2,6-difluorophenyl)piperidin-4-ol hydrochloride

  • MDL number MFCD27500987
  • Cas Number : 1803595-27-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155493

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155493
Availability details for
Quantity
  • Availability
  • Add to Cart