ENA413171407 Enamine

O-[(6-methylpyridin-2-yl)methyl]hydroxylamine dihydrochloride

  • MDL number MFCD27500989
  • Cas Number : 1803590-34-2
  • Molecular Weight : 211.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-155511

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155511
Availability details for
Quantity
  • Availability
  • Add to Cart