ENA413171409 Enamine

(5-chloro-1-methyl-1H-1,3-benzodiazol-2-yl)methanamine hydrochloride

  • MDL number MFCD27500991
  • Cas Number : 1803582-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155527
Availability details for
Quantity
  • Availability
  • Add to Cart