ENA413171412 Enamine

6-phenyl-2,3-dihydro-1H-inden-1-amine hydrochloride

  • MDL number MFCD27500994
  • Cas Number : 1803582-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155585

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155585
Availability details for
Quantity
  • Availability
  • Add to Cart