ENA413171413 Enamine

4-[2-(dimethylamino)ethenyl]benzonitrile

  • MDL number MFCD27920224
  • Cas Number : 20973-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155606

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155606
Availability details for
Quantity
  • Availability
  • Add to Cart