ENA413171414 Enamine

N-methoxy-N-methylthiane-4-carboxamide

  • MDL number MFCD27500996
  • Cas Number : 887481-46-1
  • Molecular Weight : 189.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methoxy-N-methyltetrahydro-2H-thiopyran-4-carboxamide
Inchi Code 1S/C8H15NO2S/c1-9(11-2)8(10)7-3-5-12-6-4-7/h7H,3-6H2,1-2H3
Inchi Key JICXQBVSROKJLD-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-155609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155609
Availability details for
Quantity
  • Availability
  • Add to Cart