ENA413171423 Enamine

5,5-dimethylmorpholine-3-carboxylic acid hydrochloride

  • MDL number MFCD27501006
  • Cas Number : 1796909-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-155694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-155694
Availability details for
Quantity
  • Availability
  • Add to Cart